Falling ball viscometer
Falling ball viscometer, 0,5 − 10⁵ mPa.s (cP)
Catalog # 616-3282
Supplier: HAAKE
Jump to:
Product Details & Documents
This falling ball viscometer provides an accurate way of measuring the viscosity, in transparent Newtonian liquids and gases.
Specially suited for the chemical (polymer solutions, solvents, inks), pharmaceutical (raw materials, glycerine), food (gelatin, sugar solutions) and mineral oil (oils, liquid hydrocarbons) industries.
Certifications: DIN 53015, ISO 12058
Delivery information: Supplied with six balls, instrument case, thermometer, cleaning tools, calibration sheet and instruction manual.
Specifications
- Range:0,5 − 10⁵ mPa.s (cP)
- Description:Falling ball viscometer
Accessories
Product Information
- Catalog #AvailabilityPrice
- Digital stopwatch with LCD for up to ten hours
Product Information
- Catalog #AvailabilityPrice
- Ball 2, borosilicate glass
Product Information
- Catalog #AvailabilityPrice
- Ball 1, borosilicate glass
Product Information
- Catalog #AvailabilityPrice
- Ball 5, Nickel-Iron
Product Information
- Catalog #AvailabilityPrice
- Ball 6, Nickel-Iron
Product Information
- Catalog #AvailabilityPrice
- Ball set (balls 1 to 6), delivered in a wooden caseBall set (balls 1 to 6), delivered in a wooden caseHAAK800-0182Log in for availability1 * 1 SET - 962,00 €
Product Information
- Catalog #AvailabilityPrice
- Ball 3, Nickel-Iron