Orders LinkContactUsLinkComponent
Sample containers, with screw cap
Sample containers, with screw cap
Sample containers, with screw cap
Supplier:  DELTALAB
Certificates
Sample containers, with screw cap
3 Options Available Below
Product Details & Documents
Ultra clear PP containers with PE leak proof screw cap and PS spoon.
  • Graduated in 100 ml increments
  • Frosted panel on side for easy writing
  • Good chemical and temperature resistance
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Documents
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Height
    Cap colour
    Version
    Packed
    Availability
    Price
  • 150 ml
    57 mm
    73 mm
    Red
    Non sterile
    Bulk
    Log in for availability
    Log in for availability
    1 * 450 ST
    1 * 450 ST - 242,00 €
    242,00 €
    1 * 450 ST
       242,00 €
  • 150 ml
    57 mm
    73 mm
    Red
    Non sterile
    Individually wrapped
    Log in for availability
    Log in for availability
    1 * 350 ST
    1 * 350 ST - 236,00 €
    236,00 €
    1 * 350 ST
       236,00 €
  • 150 ml
    57 mm
    73 mm
    Red
    Sterile
    Individually wrapped
    Log in for availability
    Log in for availability
    1 * 350 ST
    1 * 350 ST - 288,00 €
    288,00 €
    1 * 350 ST
       288,00 €