Pasteur pipettes
Supplier: DELTALAB
Jump to:
About this item
PE, transparent.
- Made of inert polyethylene
- Sterile models are sterilised by ethylene oxide
- Volumetric tolerance: ±10%
2 Options Available Below
Product Details & Documents
PE, transparent.
- Made of inert polyethylene
- Sterile models are sterilised by ethylene oxide
- Volumetric tolerance: ±10%
Documents
Certificates
Search by Lot NumberProduct Family Options
Product Information
- CapacityDrop vol.Bulb drawStem ØLengthSterilePackedAvailabilityPrice
- DELT210023Fine tip pipetteFine tip pipette1,5 ml20 µl1 ml3 mm104 mm+Individually wrappedLog in for availability1 * 500 ST236,00 €
- DELT200005Pasteur pipette, general purposePasteur pipette, general purpose4 ml40 µl3,1 ml2,5 mm150 mm-6×500Discontinued-
- DELT210024Fine tip pipetteFine tip pipette5,5 ml20 µl3 ml3 mm153 mm+Individually wrappedLog in for availability1 * 500 ST254,00 €