Order Entry
Belgium
ContactUsLinkComponent
 
Microplate shakers, Jitterbug
Catalog # BOEI130000-2
Supplier:  BOEKEL SCIENTIFIC
undefined
Microplate shakers, Jitterbug
Catalog # BOEI130000-2
Supplier:  BOEKEL SCIENTIFIC

Specifications

  • No. of microplates held:
    2
  • Orbit:
    1,0 mm
  • Shaking motion:
    Rotating counterweight
  • Speed range:
    575 − 1500 min⁻¹
  • Description:
    Heated microplate shaker, jitterbug
  • Model:
    Jitterbug
  • Electrical:
    230 V, 50/60 Hz
  • Weight:
    4,99 kg
  • W×D×H:
    254×254×102 mm
  • Pk:
    1

Specifications

About this item

The Jitterbug microplate shaker is ideal for immunoassays like ELISA, PCR and other research procedures requiring heating and shaking of microplates.

  • Accommodates two standard microplates (96-, 384-, 1536-well) or strip trays
  • Independently set temperature, shaking speed and mixing duration
  • Select from eight mixing speeds
  • Smooth, quiet operation
  • Excellent temperature uniformity

Includes inner lid to reduce the risk of contamination.