Orders LinkContactUsLinkComponent
Sample container, with screw cap
Sample container, with screw cap
Sample container, with screw cap
Supplier:  DELTALAB
Certificates
Sample container, with screw cap
4 Options Available Below
Product Details & Documents
PP, translucent, with screw cap.
  • Good chemical resistance
  • Good temperature resistance
Straight walled containers for sample collection, storage and transport.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Documents
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Height
    Type
    Packed
    Availability
    Price
  • 30 ml
    25 mm
    90 mm
    -
    bulk
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 126,00 €
    126,00 €
    1 * 400 ST
       126,00 €
  • 30 ml
    25 mm
    90 mm
    -
    individually wrapped
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 169,00 €
    169,00 €
    1 * 400 ST
       169,00 €
  • 30 ml
    25 mm
    90 mm
    Sterile
    individually wrapped
    Log in for availability
    Log in for availability
    1 * 400 ST
    1 * 400 ST - 212,00 €
    212,00 €
    1 * 400 ST
       212,00 €
  • 30 ml
    25 mm
    90 mm
    With spoon
    Log in for availability
    Log in for availability
    1 * 40 ST
    1 * 40 ST - 15,90 €
    15,90 €
    1 * 40 ST
       15,90 €