Orders LinkContactUsLinkComponent
Sample container, with screw cap
Sample container, with screw cap
Sample container, with screw cap
Supplier:  DELTALAB
Certificates
Sample container, with screw cap
2 Options Available Below
Product Details & Documents
Graduated containers made of PP or PS, with PE caps.
  • Good chemical resistance
  • Good temperature resistance
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904
Documents
Product Family Options
Product Information
  • Capacity
    Ext.Ø
    Height
    Colour
    Material
    Availability
    Price
  • 40 ml
    30 mm
    70 mm
    Clear
    PS
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 54,90 €
    54,90 €
    1 * 100 ST
       54,90 €
  • 40 ml
    30 mm
    70 mm
    Clear
    PP, clear
    Log in for availability
    Log in for availability
    1 * 800 ST
    1 * 800 ST - 442,00 €
    442,00 €
    1 * 800 ST
       442,00 €