Tissue processing cassettes, Histosette I
Supplier: DELTALAB
Jump to:
About this item
Made of acetal polymer. With one-piece integral lid. To lock the cassette, just snap apart the lid, turn it round and lock it into the base of the cassette.
- Writing area at 30° angle
3 Options Available Below
Product Details & Documents
Made of acetal polymer. With one-piece integral lid. To lock the cassette, just snap apart the lid, turn it round and lock it into the base of the cassette.
- Writing area at 30° angle