Orders LinkContactUsLinkComponent
PCR microtubes and strips
PCR microtubes and strips
8-strip PCR tubes, without caps, Clear, 0,2 ml, 125 tubes/pack
Catalog # 732-2463
Supplier:   BIOTIX
PCR microtubes and strips
Catalog # 732-2463
Supplier:  BIOTIX
PCR microtubes and strips
PCR microtubes and strips
8-strip PCR tubes, without caps, Clear, 0,2 ml, 125 tubes/pack
Price
Inquire For Price
Log in for availability
1 * 125 ST
Product Details & Documents
Medical grade virgin polypropylene.
  • Excellent chemical and temperature resistance
  • Biologically inert
  • BioReady™ certified free from DNase, RNase and endotoxin
Superior clarity for sample visualisation.
Specifications
  • Packed:
    125 tubes/pack
  • Capacity:
    0,2 ml
  • Colour:
    Clear
  • Description:
    8-strip PCR tubes, without caps
  • Seq:
    02