ContactUsLinkComponent
Capillary column end caps
  555-9468
undefined
Capillary column end caps
  555-9468
  • Description:
    Capillary column end caps, paired
  • Seq:
    01
  • Pk:
    10
These capillary column end caps are universal and fit any capillary column.
  • Reusable
  • Eliminate column contamination caused by leaving a column unsealed or sealed with a septum
  • Colour coded to ensure that the column is reinstalled the same way it came out