Orders LinkContactUsLinkComponent
Transport containers
Transport containers
Transport containers
Supplier:  DELTALAB
Certificates
Transport containers
Transport containers
3 Options Available Below
Product Details & Documents
Designed for slide transport and protection.
  • Made of polypropylene
  • Easy snap-lock
  • Not indicated for liquid samples
Compartment shape ensures tight fit: no risk of damaging slides and to send by mail, recommend to use special envelopes.
Documents
Product Family Options
Product Information
  • Material
    W×D×H
    No. of slides
    Availability
    Price
  • Slide mailers
    PP
    87×47×16 mm
    2
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 56,40 €
    56,40 €
    1 * 100 ST
       56,40 €
  • DELT979930
    Slide mailers
    Slide mailers
    PP
    84×71×6 mm
    2
    Log in for availability
    Log in for availability
    1 * 250 ST
    1 * 250 ST - 279,00 €
    279,00 €
    1 * 250 ST
       279,00 €
  • Slide mailers
    PP
    82×17×29 mm
    5
    Log in for availability
    Log in for availability
    1 * 10 ST
    1 * 10 ST - 9,60 €
    9,60 €
    1 * 10 ST
       9,60 €