Orders LinkContactUsLinkComponent
Ring weights for flasks
Supplier:   DELTALAB
Ring weights for flasks
Supplier:  DELTALAB
Certificates
Ring weights for flasks
3 Options Available Below
Product Details & Documents
Closed flask weight rings with heavy vinyl coating, which is ideal to improve the stability of Erlenmeyer flasks in a water bath.
  • High chemical resistance
Product Family Options
Product Information
  • For
    Ø int.
    Availability
    Price
  • DELT192470
    Viny coated lead ring weight
    Viny coated lead ring weight
    Erlenmeyer 125 - 500 ml
    48 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 131,00 €
    131,00 €
    1 * 1 ST
       131,00 €
  • DELT192471
    Viny coated lead ring weight
    Viny coated lead ring weight
    Erlenmeyer 250 - 1000 ml
    51 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 143,00 €
    143,00 €
    1 * 1 ST
       143,00 €
  • DELT192472
    Viny coated lead ring weight
    Viny coated lead ring weight
    Erlenmeyer 500 - 2000 ml
    57 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 143,00 €
    143,00 €
    1 * 1 ST
       143,00 €