ContactUsLinkComponent
Blue light LED transilluminators, Syngene UltraSlim
Blue light LED transilluminators, Syngene UltraSlim
Catalog # SYNOGEULTRASLIMLED
undefined
Blue light LED transilluminators, Syngene UltraSlim
Catalog # SYNOGEULTRASLIMLED
Specifications
  • Description:
    Blue light LED transilluminator, for InGenius3 and U:Genius3 systems (100×120 mm, 470 nm)
  • Dimensions:
    210×210×30 mm
  • Seq:
    01
  • Pk:
    1
Specifications
About this item
UltraSlim-LED transilluminators for superior gel illumination.
  • Compact
  • Even blue epi LED illumination - high sensitivity with improved S/N ratio
  • Image a range of fluorophores, eg. EtBr, SYBR®Safe, GelGreen, SYBR®Gold, SYBR®Green, SYPRO®Ruby, SYPRO®Orange, GelRed™
  • LED is safe to skin and eyes with no damage to DNA