Fluid aspiration systems, WOB-L®
These heavy-duty diaphragm pumps are specially designed for use in suction and filtration.
The connections with the pump are made by means of a quick plug.
: System includes an autoclavable vessel, adapters, tube, hydrophobic filters, control vacuum and manometer.
- Catalog No:
- ILMVU2511C-75
- ILMVU2515C-75
- Modèle:
- WOB-L® 2511
- WOB-L® 2515
- Débit:
- 9200 (at 50 Hz)
11000 (at 60 Hz) - 28000 (at 50 Hz)
34000 (at 60 Hz)
- Niveau de bruit:
- 45 dB(A)
- 46 dB(A)
Frequently Bought Together