Orders LinkContactUsLinkComponent
pH Buffer solutions, CX-1
pH Buffer solutions, CX-1
pH Buffer solutions, CX-1
pH Buffer solutions, CX-1
pH Buffer solutions, CX-1
Delivers robust reproducible gradients with the Thermo Scientific™ pH buffer platform that is applicable to a wide range of MAbs and ready to use with existing LC columns and systems, without the need to formulate mobile phases.
Product Family Options
  • Description
    pH value
    Pk
  • pH gradient buffer, CX-1, buffer A
    5,6
    125 mL
    1 * 125 mL
    1 * 125 mL - 217,00 €
    217,00 €
    1 * 125 mL
       217,00 €
  • pH gradient buffer, CX-1, buffer A
    5,6
    250 mL
    1 * 250 mL
    1 * 250 mL - 348,00 €
    348,00 €
    1 * 250 mL
       348,00 €
  • pH gradient buffer, CX-1, buffer B
    10,2
    125 mL
    1 * 125 mL
    1 * 125 mL - 217,00 €
    217,00 €
    1 * 125 mL
       217,00 €
  • pH gradient buffer, CX-1, buffer B
    10,2
    250 mL
    1 * 250 mL
    1 * 250 mL - 348,00 €
    348,00 €
    1 * 250 mL
       348,00 €