ContactUsLinkComponent
Measuring beakers with handle, PP
  215-4360
 :  LAMAPLAST
 : 1161
undefined
Measuring beakers with handle, PP
  215-4360
 :  LAMAPLAST
 : 1161
Select an Option
65 mm
Select an Option
250 ml
Select an Option
95 mm
Select an Option
01
Select an Option
1
PP, translucent.
  • Good chemical resistance
  • Resistant to heat up to +121 °C (for 20 min), prolonged use +100 °C
  • Smooth surfaces for easy, fast cleaning
Ideal for general laboratory use and a wide variety of other applications. With embossed scale and spout.
Frequently Bought Together