Orders LinkContactUsLinkComponent
Precision hotplate, PZ
Precision hotplate, PZ
Supplier:   GESTIGKEIT HARRY
Precision hotplate, PZ
Supplier:  GESTIGKEIT HARRY
Certificates
Precision hotplate, PZ
5 Options Available Below
Product Details & Documents
Precision hotplates are equipped with adjustable temperature sensors +5 to +300 °C (PZ28-1 +30 to +110 °C). These sensors function as an emergency control which protect both the appliance and the substance to be heated against dangerous overheating.
Product Family Options
Product Information
  • Heating area
    Temp.
    Heating capacity
    Plate size
    Type
    Availability
    Price
  • 200×280 mm
    20...99,9 °C
    500 W
    210×300 mm
    PZ 28-1
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 230,00 €
    2 230,00 €
    1 * 1 ST
       2 230,00 €
  • 200×280 mm
    20...300 °C
    1100 W
    210×300 mm
    PZ 28-2
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 230,00 €
    2 230,00 €
    1 * 1 ST
       2 230,00 €
  • 350×350 mm
    20...300 °C
    2200 W
    365×365 mm
    PZ 35
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 750,00 €
    2 750,00 €
    1 * 1 ST
       2 750,00 €
  • 610×160 mm
    20...300 °C
    2000 W
    620×200 mm
    PZ 60
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 2 690,00 €
    2 690,00 €
    1 * 1 ST
       2 690,00 €
  • 430×580 mm
    20...300 °C
    3300 W
    430×580 mm
    PZ 72
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 3 380,00 €
    3 380,00 €
    1 * 1 ST
       3 380,00 €