Orders LinkContactUsLinkComponent
Counting chamber with grid
Counting chamber with grid
Counting chambers with grid, 20×20 mm
Catalog # DIVEGRID-1000
Supplier:   DIVERSIFIED BIOTECH
Counting chamber with grid
Catalog # DIVEGRID-1000
Counting chamber with grid
Counting chambers with grid, 20×20 mm
Price
94,00 €
Log in for availability
1 * 40 ST
Product Details & Documents
Disposable polyester with acrylic adhesive grid stickers for micro or semi-micro objects.
  • Inert and stable under most conditions
  • Compatible with different kinds of microscopes
  • Separate coverslips can be placed over each sticker
These slide grids are either used with dry or suspended specimens, such as cells held in buffer, under cover slips.
Specifications
  • W×D:
    20×20 mm
  • Description:
    Counting chambers with grid