Orders LinkContactUsLinkComponent
Immersion probe
Immersion probe
Immersion probe, NiCr-Ni, class 1, flexible jacket material (inconel), electrically isolated, −200…+1150 °C, 1,5×130 mm
Catalog # 620-4707
Supplier:   Greisinger
Immersion probe
Catalog # 620-4707
Supplier:  Greisinger
Immersion probe
Immersion probe
Immersion probe, NiCr-Ni, class 1, flexible jacket material (inconel), electrically isolated, −200…+1150 °C, 1,5×130 mm
Price
Inquire For Price
Log in for availability
1 * 1 ST
Product Details & Documents
Specifications
  • Range:
    −200…+1150 °C
  • Description:
    Immersion probe, NiCr-Ni, class 1, flexible jacket material (inconel), electrically isolated
  • Ø×L:
    1,5×130 mm