Order Entry
Belgium
Orders LinkContactUsLinkComponent
Heating tapes

Heating tapes

Heating tape with 1,5 m length, 525 W, 1,5 m, 525 W
Catalog # WINKWBQ00901500525
Supplier:   WINKLER
Heating tapes
Catalog # WINKWBQ00901500525
Supplier:  WINKLER
Heating tapes
Heating tapes

Heating tape with 1,5 m length, 525 W, 1,5 m, 525 W

Price
361,53 €
Log in for availability
Product Details & Documents
Specifications
  • Description:
    Heating tape with 1,5 m length, 525 W
  • Length:
    1,5 m
  • Output:
    525 W
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.