Saisie de commande
Belgium
Lien pour commandesContactUsLinkComponent
Heating Cords
Supplier:   WINKLER
Heating Cords
Supplier:  WINKLER
Certificates
Heating Cords
9 Options Available Below
Product Details & Documents
Heating cord which consists of one heating spiral insulated with glass-silk.
  • With flexible glass-silk outer braiding
  • Max. temperature 400 °C
Product Family Options
Product Information