ContactUsLinkComponent
Adhesion slides, Apex Superior
Adhesion slides, Apex Superior
Catalog # 630-2184
Supplier:  LEICA HISTOLOGY
Adhesion slides, Apex Superior
Catalog # 630-2184
Supplier:  LEICA HISTOLOGY
Specifications
  • W×D:
    76×26 mm
  • Colour:
    White
  • Description:
    Adhesion slides
  • Pk:
    72
Specifications
About this item
These adhesive slides are coated for a positive charge to enhance hydrophilicity.
  • Homogeneous covering by aqueous stains and reagents
  • Consistent full slide staining
For demanding procedures such as standard IHC and HIER techniques.