Orders LinkContactUsLinkComponent
Temperature indicating tubes
Temperature indicating tubes
Temperature indicating tubes
Certificates
Temperature indicating tubes
Temperature indicating tubes
3 Options Available Below
Product Details & Documents
Temperature indicating tubes are used for autoclaves and hot air sterilisers.
Sterilisation is detected by the colour change of the control tube from red to green. The tubes should be placed as close as possible next to or in the load.
Product Family Options
Product Information
  • Availability
    Price
  • KOEHK-1250
    For autoclaves up to 125 °C
    For autoclaves up to 125 °C
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 241,00 €
    241,00 €
    1 * 1 ST
       241,00 €
  • 391-0085
    For hot-air sterilization 180 °C/12 min
    For hot-air sterilization 180 °C/12 min
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 188,00 €
    188,00 €
    1 * 100 ST
       188,00 €
  • 391-0084
    For hot-air sterilization 125 °C/15 min
    For hot-air sterilization 125 °C/15 min
    Log in for availability
    Log in for availability
    1 * 100 ST
    1 * 100 ST - 255,00 €
    255,00 €
    1 * 100 ST
       255,00 €