Orders LinkContactUsLinkComponent
Density hydrometers
Density hydrometers
Supplier:   Amarell
Density hydrometers
Supplier:  Amarell
Certificates
Density hydrometers
Density hydrometers
87 Options Available Below
Product Details & Documents
General purpose hydrometers for density areas.
  • Paper scale and shot weighted body
  • Hydrometer accuracy ±1 scale division
  • Thermometer inside the hydrometer body
Used in laboratories and industry.
Product Family Options
Product Information
  • Range
    Division
    Length
    Availability
    Price
  • AMARH800004
    Hydrometer 1
    Hydrometer 1
    0,700 - 0,760 g/cm³
    0,001 g/cm³
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 29,10 €
    29,10 €
    1 * 1 ST
       29,10 €
  • AMARH800006
    Hydrometer 2
    Hydrometer 2
    0,760 - 0,820 g/cm³
    0,001 g/cm³
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 29,10 €
    29,10 €
    1 * 1 ST
       29,10 €
  • AMARH800008
    Hydrometer 3
    Hydrometer 3
    0,820 - 0,880 g/cm³
    0,001 g/cm³
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 29,10 €
    29,10 €
    1 * 1 ST
       29,10 €
  • AMARH800010
    Hydrometer 4
    Hydrometer 4
    0,880 - 0,940 g/cm³
    0,001 g/cm³
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 29,10 €
    29,10 €
    1 * 1 ST
       29,10 €
  • AMARH800012
    Hydrometer 5
    Hydrometer 5
    0,940 - 1,000 g/cm³
    0,001 g/cm³
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 29,10 €
    29,10 €
    1 * 1 ST
       29,10 €
  • AMARH800014
    Hydrometer 6
    Hydrometer 6
    1,000 - 1,060 g/cm³
    0,001 g/cm³
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,30 €
    31,30 €
    1 * 1 ST
       31,30 €
  • AMARH800016
    Hydrometer 7
    Hydrometer 7
    1,060 - 1,120 g/cm³
    0,001 g/cm³
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,30 €
    31,30 €
    1 * 1 ST
       31,30 €
  • AMARH800018
    Hydrometer 8
    Hydrometer 8
    1,120 - 1,180 g/cm³
    0,001 g/cm³
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,30 €
    31,30 €
    1 * 1 ST
       31,30 €
  • AMARH800020
    Hydrometer 9
    Hydrometer 9
    1,180 - 1,240 g/cm³
    0,001 g/cm³
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,30 €
    31,30 €
    1 * 1 ST
       31,30 €
  • AMARH800022
    Hydrometer 10
    Hydrometer 10
    1,240 - 1,300 g/cm³
    0,001 g/cm³
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,30 €
    31,30 €
    1 * 1 ST
       31,30 €
  • AMARH800024
    Hydrometer 11
    Hydrometer 11
    1,300 - 1,360 g/cm³
    0,001 g/cm³
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 33,80 €
    33,80 €
    1 * 1 ST
       33,80 €
  • AMARH800026
    Hydrometer 12
    Hydrometer 12
    1,360 - 1,420 g/cm³
    0,001 g/cm³
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 33,80 €
    33,80 €
    1 * 1 ST
       33,80 €
  • AMARH800028
    Hydrometer 13
    Hydrometer 13
    1,420 - 1,480 g/cm³
    0,001 g/cm³
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 33,80 €
    33,80 €
    1 * 1 ST
       33,80 €
  • AMARH800030
    Hydrometer 14
    Hydrometer 14
    1,480 - 1,540 g/cm³
    0,001 g/cm³
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 33,80 €
    33,80 €
    1 * 1 ST
       33,80 €
  • AMARH800032
    Hydrometer 15
    Hydrometer 15
    1,540 - 1,600 g/cm³
    0,001 g/cm³
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 33,80 €
    33,80 €
    1 * 1 ST
       33,80 €
  • AMARH800034
    Hydrometer 16
    Hydrometer 16
    1,600 - 1,660 g/cm³
    0,001 g/cm³
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,30 €
    38,30 €
    1 * 1 ST
       38,30 €
  • AMARH800036
    Hydrometer 17
    Hydrometer 17
    1,660 - 1,720 g/cm³
    0,001 g/cm³
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,30 €
    38,30 €
    1 * 1 ST
       38,30 €
  • AMARH800038
    Hydrometer 18
    Hydrometer 18
    1,720 - 1,780 g/cm³
    0,001 g/cm³
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,30 €
    38,30 €
    1 * 1 ST
       38,30 €
  • AMARH800040
    Hydrometer 19
    Hydrometer 19
    1,780 - 1,840 g/cm³
    0,001 g/cm³
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,30 €
    38,30 €
    1 * 1 ST
       38,30 €
  • AMARH800042
    Hydrometer 20
    Hydrometer 20
    1,840 - 1,900 g/cm³
    0,001 g/cm³
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,30 €
    38,30 €
    1 * 1 ST
       38,30 €
  • AMARH800044
    Hydrometer 21
    Hydrometer 21
    1,900 - 1,960 g/cm³
    0,001 g/cm³
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,30 €
    38,30 €
    1 * 1 ST
       38,30 €
  • AMARH800046
    Hydrometer 22
    Hydrometer 22
    1,960 - 2,020 g/cm³
    0,001 g/cm³
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,30 €
    38,30 €
    1 * 1 ST
       38,30 €
  • AMARH800130
    Hydrometer 00
    Hydrometer 00
    0,600 - 0,660 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,30 €
    51,30 €
    1 * 1 ST
       51,30 €
  • AMARH800132/WC
    Hydrometer 0 with WC certificate
    Hydrometer 0 with WC certificate
    0,650 - 0,710 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • AMARH800132
    Hydrometer 0
    Hydrometer 0
    0,650 - 0,710 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,30 €
    51,30 €
    1 * 1 ST
       51,30 €
  • AMARH800134/WC
    Hydrometer 1 with WC certificate
    Hydrometer 1 with WC certificate
    0,700 - 0,760 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • AMARH800134
    Hydrometer 1
    Hydrometer 1
    0,700 - 0,760 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,30 €
    51,30 €
    1 * 1 ST
       51,30 €
  • AMARH800136/WC
    Hydrometer 2 with WC certificate
    Hydrometer 2 with WC certificate
    0,760 - 0,820 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • AMARH800136
    Hydrometer 2
    Hydrometer 2
    0,760 - 0,820 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,30 €
    51,30 €
    1 * 1 ST
       51,30 €
  • AMARH800138
    Hydrometer 3
    Hydrometer 3
    0,820 - 0,880 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,30 €
    51,30 €
    1 * 1 ST
       51,30 €
  • AMARH800140/WC
    Hydrometer 4 with WC certificate
    Hydrometer 4 with WC certificate
    0,880 - 0,940 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • AMARH800140
    Hydrometer 4
    Hydrometer 4
    0,880 - 0,940 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,30 €
    51,30 €
    1 * 1 ST
       51,30 €
  • AMARH800142/WC
    Hydrometer 5 with WC certificate
    Hydrometer 5 with WC certificate
    0,940 - 1,000 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST
  • AMARH800142
    Hydrometer 5
    Hydrometer 5
    0,940 - 1,000 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 51,30 €
    51,30 €
    1 * 1 ST
       51,30 €
  • AMARH800144
    Hydrometer 6
    Hydrometer 6
    1,000 - 1,060 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,20 €
    34,20 €
    1 * 1 ST
       34,20 €
  • AMARH800146
    Hydrometer 7
    Hydrometer 7
    1,060 - 1,120 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,20 €
    34,20 €
    1 * 1 ST
       34,20 €
  • AMARH800148
    Hydrometer 8
    Hydrometer 8
    1,120 - 1,180 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,20 €
    34,20 €
    1 * 1 ST
       34,20 €
  • AMARH800150
    Hydrometer 9
    Hydrometer 9
    1,180 - 1,240 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,20 €
    34,20 €
    1 * 1 ST
       34,20 €
  • 614-0123
    Hydrometer 10
    Hydrometer 10
    1,240 - 1,300 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 34,20 €
    34,20 €
    1 * 1 ST
       34,20 €
  • AMARH800154
    Hydrometer 11
    Hydrometer 11
    1,300 - 1,360 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 59,50 €
    59,50 €
    1 * 1 ST
       59,50 €
  • AMARH800156
    Hydrometer 12
    Hydrometer 12
    1,360 - 1,420 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 59,50 €
    59,50 €
    1 * 1 ST
       59,50 €
  • AMARH800158
    Hydrometer 13
    Hydrometer 13
    1,420 - 1,480 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 59,50 €
    59,50 €
    1 * 1 ST
       59,50 €
  • AMARH800160
    Hydrometer 14
    Hydrometer 14
    1,480 - 1,540 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 59,50 €
    59,50 €
    1 * 1 ST
       59,50 €
  • AMARH800162
    Hydrometer 15
    Hydrometer 15
    1,540 - 1,600 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 59,50 €
    59,50 €
    1 * 1 ST
       59,50 €
  • AMARH800164
    Hydrometer 16
    Hydrometer 16
    1,600 - 1,660 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 62,90 €
    62,90 €
    1 * 1 ST
       62,90 €
  • AMARH800166
    Hydrometer 17
    Hydrometer 17
    1,660 - 1,720 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 62,90 €
    62,90 €
    1 * 1 ST
       62,90 €
  • AMARH800168
    Hydrometer 18
    Hydrometer 18
    1,720 - 1,780 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 62,90 €
    62,90 €
    1 * 1 ST
       62,90 €
  • AMARH800170
    Hydrometer 19
    Hydrometer 19
    1,780 - 1,840 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 62,90 €
    62,90 €
    1 * 1 ST
       62,90 €
  • AMARH800172
    Hydrometer 20
    Hydrometer 20
    1,840 - 1,900 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 62,90 €
    62,90 €
    1 * 1 ST
       62,90 €
  • AMARH800174
    Hydrometer 21
    Hydrometer 21
    1,900 - 1,960 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 62,90 €
    62,90 €
    1 * 1 ST
       62,90 €
  • AMARH800176
    Hydrometer 21
    Hydrometer 21
    1,960 - 2,020 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 62,90 €
    62,90 €
    1 * 1 ST
       62,90 €
  • AMARH801050
    Hydrometer
    Hydrometer
    0,600 - 0,700 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,30 €
    22,30 €
    1 * 1 ST
       22,30 €
  • AMARH801052
    Hydrometer
    Hydrometer
    0,700 - 0,800 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,10 €
    18,10 €
    1 * 1 ST
       18,10 €
  • AMARH801054
    Hydrometer
    Hydrometer
    0,800 - 0,900 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,10 €
    18,10 €
    1 * 1 ST
       18,10 €
  • AMARH801056
    Hydrometer
    Hydrometer
    0,900 - 1,000 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,10 €
    18,10 €
    1 * 1 ST
       18,10 €
  • AMARH801058
    Hydrometer
    Hydrometer
    1,000 - 1,100 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 19,70 €
    19,70 €
    1 * 1 ST
       19,70 €
  • AMARH801060
    Hydrometer
    Hydrometer
    1,100 - 1,200 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 21,40 €
    21,40 €
    1 * 1 ST
       21,40 €
  • AMARH801062
    Hydrometer
    Hydrometer
    1,200 - 1,300 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,50 €
    22,50 €
    1 * 1 ST
       22,50 €
  • AMARH801064
    Hydrometer
    Hydrometer
    1,300 - 1,400 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,30 €
    22,30 €
    1 * 1 ST
       22,30 €
  • AMARH801066
    Hydrometer
    Hydrometer
    1,400 - 1,500 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,10 €
    24,10 €
    1 * 1 ST
       24,10 €
  • 614-0124
    Hydrometer
    Hydrometer
    1,500 - 1,600 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,10 €
    24,10 €
    1 * 1 ST
       24,10 €
  • AMARH801070
    Hydrometer
    Hydrometer
    1,600 - 1,700 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 31,80 €
    31,80 €
    1 * 1 ST
       31,80 €
  • AMARH801072
    Hydrometer
    Hydrometer
    1,700 - 1,800 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 32,20 €
    32,20 €
    1 * 1 ST
       32,20 €
  • AMARH801074
    Hydrometer
    Hydrometer
    1,800 - 1,900 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,10 €
    26,10 €
    1 * 1 ST
       26,10 €
  • AMARH801076
    Hydrometer
    Hydrometer
    1,900 - 2,000 g/cm³
    0,001 g/cm³
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 39,40 €
    39,40 €
    1 * 1 ST
       39,40 €
  • AMARH801282
    Hydrometer
    Hydrometer
    0,850 - 1,000 g/cm³
    0,005 g/cm³
    180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 42,50 €
    42,50 €
    1 * 1 ST
       42,50 €
  • AMARH801284
    Hydrometer
    Hydrometer
    1,000 - 1,250 g/cm³
    0,005 g/cm³
    180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 45,20 €
    45,20 €
    1 * 1 ST
       45,20 €
  • AMARH801286
    Hydrometer
    Hydrometer
    1,250 - 1,500 g/cm³
    0,005 g/cm³
    180 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 45,20 €
    45,20 €
    1 * 1 ST
       45,20 €
  • AMARH801360
    Hydrometer
    Hydrometer
    0,600 - 0,800 g/cm³
    0,002 g/cm³
    280 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,60 €
    18,60 €
    1 * 1 ST
       18,60 €
  • AMARH801362
    Hydrometer
    Hydrometer
    0,800 - 1,000 g/cm³
    0,002 g/cm³
    280 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 39,00 €
    39,00 €
    1 * 1 ST
       39,00 €
  • AMARH801364
    Hydrometer
    Hydrometer
    1,000 - 1,200 g/cm³
    0,002 g/cm³
    280 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,90 €
    23,90 €
    1 * 1 ST
       23,90 €
  • AMARH801366
    Hydrometer
    Hydrometer
    1,200 - 1,400 g/cm³
    0,002 g/cm³
    280 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 23,90 €
    23,90 €
    1 * 1 ST
       23,90 €
  • AMARH801368
    Hydrometer
    Hydrometer
    1,400 - 1,600 g/cm³
    0,002 g/cm³
    280 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 25,20 €
    25,20 €
    1 * 1 ST
       25,20 €
  • AMARH801370
    Hydrometer
    Hydrometer
    1,600 - 1,800 g/cm³
    0,002 g/cm³
    280 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 25,70 €
    25,70 €
    1 * 1 ST
       25,70 €
  • AMARH801372
    Hydrometer
    Hydrometer
    1,800 - 2,000 g/cm³
    0,002 g/cm³
    280 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,10 €
    26,10 €
    1 * 1 ST
       26,10 €
  • AMARH801620
    Hydrometer
    Hydrometer
    0,700 - 1,000 g/cm³
    0,005 g/cm³
    280 - 300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 18,20 €
    18,20 €
    1 * 1 ST
       18,20 €
  • 614-0125
    Hydrometer
    Hydrometer
    1,000 - 1,300 g/cm³
    0,005 g/cm³
    280 - 300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,40 €
    20,40 €
    1 * 1 ST
       20,40 €
  • AMARH801624
    Hydrometer
    Hydrometer
    1,000 - 1,500 g/cm³
    0,005 g/cm³
    280 - 300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 20,60 €
    20,60 €
    1 * 1 ST
       20,60 €
  • 614-0126
    Hydrometer
    Hydrometer
    1,300 - 1,600 g/cm³
    0,005 g/cm³
    280 - 300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 27,90 €
    27,90 €
    1 * 1 ST
       27,90 €
  • AMARH801628
    Hydrometer
    Hydrometer
    1,500 - 2,000 g/cm³
    0,005 g/cm³
    280 - 300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 24,10 €
    24,10 €
    1 * 1 ST
       24,10 €
  • 614-0121
    Hydrometer
    Hydrometer
    2,000 - 2,500 g/cm³
    0,005 g/cm³
    280 - 300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 89,90 €
    89,90 €
    1 * 1 ST
       89,90 €
  • AMARH801632
    Hydrometer
    Hydrometer
    2,500 - 3,000 g/cm³
    0,005 g/cm³
    280 - 300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 89,90 €
    89,90 €
    1 * 1 ST
       89,90 €
  • 614-0531
    Hydrometer
    Hydrometer
    0,700 - 2,000 g/cm³
    0,02 g/cm³
    280 - 350 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 38,10 €
    38,10 €
    1 * 1 ST
       38,10 €
  • AMARH801804
    Hydrometer
    Hydrometer
    1,000 - 2,000 g/cm³
    0,01 g/cm³
    280 - 350 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 16,70 €
    16,70 €
    1 * 1 ST
       16,70 €
Range finder
Product Information
  • Range
    Division
    Length
    Availability
    Price
  • AMARH800180
    Range finder for the hydrometers 00 - 22
    Range finder for the hydrometers 00 - 22
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 98,60 €
    98,60 €
    1 * 1 ST
       98,60 €
Complete set
Product Information
  • Range
    Division
    Length
    Availability
    Price
  • AMARH800060
    Completely set with 25 instruments (00 to 22, incl. range finder)
    Completely set with 25 instruments (00 to 22, incl. range finder)
    160 mm
    Log in for availability
    Log in for availability
    1 * 1 SET
    1 * 1 SET
  • AMARH800190
    Completely set with 25 instruments (00 to 22, incl. range finder)
    Completely set with 25 instruments (00 to 22, incl. range finder)
    300 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST