Orders LinkContactUsLinkComponent
Slide boxes
Slide boxes
Slide box, Green, PS, 100, 210×166×30 mm
Catalog # KOEHK-2451
Slide boxes
Catalog # KOEHK-2451
Slide boxes
Slide boxes
Slide box, Green, PS, 100, 210×166×30 mm
Price
17,10 €
Log in for availability
1 * 1 ST
Product Details & Documents
Green polystyrene slide box for 100 microscope slides.
  • Coloured
  • Stackable with dustproof hinged lid
  • Interlocked and index card
Specifications
  • No. of slides:
    100
  • Colour:
    Green
  • Description:
    Slide box
  • Material:
    PS
  • Seq:
    01
  • W×D×H:
    210×166×30 mm