ContactUsLinkComponent
Bench top protector sheets, KIMTECH SCIENCE*
Bench top protector sheets, KIMTECH SCIENCE*
Catalog # KIMB75460
Supplier:  KIMBERLY CLARK
undefined
Bench top protector sheets, KIMTECH SCIENCE*
Catalog # KIMB75460
Supplier:  KIMBERLY CLARK
Specifications
  • Packed:
    Sheet
  • Colour:
    Green
  • L×W:
    457×495 mm
  • Seq:
    01
  • Pk:
    50
Specifications
About this item
This advanced bench top protector made from Airlaid material features a polyethylene backing for excellent slip resistance.
  • Quickly absorbs spills
  • Flat, stable surface helps protect experiments and delicate glassware
  • Remains flat, even after drying