Orders LinkContactUsLinkComponent
Sample Bags
Sample Bags
Sampling bags, sodium thiosulphate pill (10 mg), 100 ml
Catalog # HACH2075333
Supplier:   Hach
Sample Bags
Catalog # HACH2075333
Supplier:  Hach
Sample Bags
Sample Bags
Sampling bags, sodium thiosulphate pill (10 mg), 100 ml
Price
52,90 €
Log in for availability
1 * 100 ST
Product Details & Documents
They are transparent PE bags that are specially designed for chlorinated water samples and also for testing recreational water, waste water and drinking water.
  • Sterile and disposable bags
  • With 'Whirl-Pak' closure
  • Write-on strip for waterproof ink
Certifications: Complies with USEPA.
Specifications
  • Capacity:
    100 ml
  • Description:
    Sampling bags, sodium thiosulphate pill (10 mg)