Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
0,00 €
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Belgium
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Home
Tubing
Transfer Tubing
Tubing, latex rubber
Tubing, latex rubber
Tubing, latex rubber
Catalog # HACH4208600
Supplier:
Hach
Expand image
Tubing, latex rubber
Catalog # HACH4208600
Supplier:
Hach
Specifications
Tubing ext.Ø:
12,7 mm
Tubing int.Ø:
9,53 mm
Wall thickness:
1,59 mm
Colour:
Translucent
Pk:
30
Specifications
About this item
Product Details
Latex, translucent.
Product Details