ContactUsLinkComponent
Replicating tool, 96-well, Bel-Blotter™, SP Bel-Art
Replicating tool, 96-well, Bel-Blotter™, SP Bel-Art
Catalog # BELAF378760002
undefined
Replicating tool, 96-well, Bel-Blotter™, SP Bel-Art
Catalog # BELAF378760002
Specifications
  • Description:
    Bel-Blotter™ replicating tool, 96-well
  • Pk:
    1
Specifications
About this item
The 96 'open-ended' pipette tips in this replicating tool are compatible with all kinds of plates and tubes.
  • Fits plates with flat, V- or U-bottom wells
  • Up to 10 µl of fluid per tip aspirated
  • Sturdy, durable polypropylene construction
  • Re-usable and steam sterilisable