ContactUsLinkComponent
Sampling swabs, NRSII™ Transwab®
Sampling swabs, NRSII™ Transwab®
Catalog # MEDWMW776
Sampling swabs, NRSII™ Transwab®
Catalog # MEDWMW776
Specifications
  • Description:
    Swabs with 10 ml buffered peptone water
  • Seq:
    002
  • Cap colour:
    Blue
  • Pk:
    500
Specifications
About this item
Preimmersed swabs for use with a self standing tube and a high visibility blue cap and swab, which is easy to find and retrive if accidentally dropped.
  • Blue cap and swab which can be easily retrived if dropped
  • Reduced packaging and storage space, ready to use
  • Ambient storage and transport
  • Precise fill for accurate assessment of contamination levels
  • Neutralisers to allow for the accurate recovery of residual bacteria on food contact surfaces
Specially recommended for sampling previously cleaned surfaces.
Certifications: ISO 18593 and other national and international standards.
Caution: *The product(s) on this page are outside the scope of the EU Directive 2019/904