Order Entry
Belgium
ContactUsLinkComponent
Blotting membranes, Amersham™ Hybond™ SEQ 0,2 PVDF
Blotting membranes, Amersham™ Hybond™ SEQ 0,2 PVDF
Catalog # 10600030.
Supplier:  Cytiva
undefined
Blotting membranes, Amersham™ Hybond™ SEQ 0,2 PVDF
Catalog # 10600030.
Supplier:  Cytiva

Specifications

  • Description:
    PVDF blotting membranes, roll, pore size 0,2 µm
  • Dimensions:
    260 mm × 4 m
  • Seq:
    01
  • Pk:
    1

Specifications

About this item

Amersham™ Hybond™ SEQ 0.2 PVDF is a 0,2 µm pore size hydrophobic membrane designed specifically for protein sequencing applications. The small pore size of this membrane minimises ‘blow-through’ and increases protein binding over a wide range of molecular weights.

  • Efficient capture of proteins during transfer minimises sample loss
  • High protein retention even after harsh washing steps
  • 0,2 µm pore size for higher surface area, resulting in better binding of low molecular weight proteins
  • Protein binding capacity over 200 µg/cm² for easy signal detection
  • Available in popular precut sizes
  • Chemical resistance needed for N-terminal sequencing