pH electrodes for fast results, InLab®
Supplier: Mettler - Toledo
Certificates
About this item
pH electrodes with S7, BNC or MultiPin™ (pH/temperature) connector.
- Liquid filled electrodes for efficient pH measurements in daily laboratory use of a wide application range
- ARGENTHAL™ reference system with silver-ion trap so that there is no risk of contamination by sulphides and proteins
- Conform to RoHS ('Restriction of Hazardous Substances' in electrical and electronic equipment) and WEEE ('Waste Electrical and Electronic Equipment')
- InLab® Versatile Pro features a replaceable ceramic junction which offers an economic alternative to open or movable sleeve junctions
Junction: Movable glass sleeve
* Integrated temperature probe (NTC30)
** Integrated temperature probe (Pt1000)
Electrolyte: 3 mol/l KCl
Ordering information: Supplied with test certificate and with (BNC connection) or without cable (S7 and MultiPin™ connections). Junction tool and set of junctions for replacing the junction of the InLab® Versatile Pro electrode are available separately.
Related Information
Certificates
Search by Lot NumberMETT51343056_TDS_EN
Datasheet
Mettler InLab® Sensors
Product Literature
Frequently Bought Together