ContactUsLinkComponent
Isofreeze racks with temperature indicator
Isofreeze racks with temperature indicator
Catalog # DELTM-572
Supplier:  DELTALAB
undefined
Isofreeze racks with temperature indicator
Catalog # DELTM-572
Supplier:  DELTALAB
Specifications
  • No. of Holes:
    96
  • Description:
    Isofreeze racks, PP, yellow-green, with temperature indicator
  • W×D×H:
    141×99×44 mm
  • Pk:
    2
Specifications
About this item
PP, with clear lid.
  • Filled with a non toxic gel that maintains 4 °C temperature for almost four hours when the rack is removed from the freezer
  • A colour indicator begins to change from green to yellow as the temperature rises above 0 degrees