Order Entry
Belgium
Orders LinkContactUsLinkComponent
Mega tissue processing cassettes

Mega tissue processing cassettes

White
Catalog # DELT44125
Supplier:   DELTALAB
Mega tissue processing cassettes
Catalog # DELT44125
Supplier:  DELTALAB
Mega tissue processing cassettes

White

Price
209,00 €
Log in for availability
1 * 500 ST
Product Details & Documents

Made from polyacetyle. Designed for larger tissue samples.

  • With an integral lid
  • Dimension: 27,4×40,5×8,15 mm
  • Holes:1×5 mm
Specifications
  • Colour:
    White