Orders LinkContactUsLinkComponent
Inserts for storage tubes, 2D Barcoded, Matrix®
Inserts for storage tubes, 2D Barcoded, Matrix®
   Sustainable Options Available
Inserts for storage tubes, 2D Barcoded, Matrix®
   Sustainable Options Available
Certificates
Inserts for storage tubes, 2D Barcoded, Matrix®
2 Options Available Below
Product Details & Documents
Color inserts snap into cap for a secure fit.
  • Inserts are removable
  • Free of DNA, DNAse, RNAse, PCR Inhibitors, endotoxins and cytotoxins
Specifications for Product Family
Specifications
  • Catalog No:
  • 479-1344
  • 479-1343
  • Environmentally Preferable:
  • Low Manufacturing Impact
  • Low Manufacturing Impact
  • Environmental certification:
  • NAR Certificate of Retirement
  • NAR Certificate of Retirement
Specifications
Product Family Options
Product Information
  • Colour
    Availability
    Price
  • 479-1344
    Insert for 0,5 ml and 1,0 ml screw top tube caps
    Insert for 0,5 ml and 1,0 ml screw top tube caps
    Purple
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 26,50 €
    26,50 €
    1 * 500 ST
       26,50 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    479-1344
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact
  • 479-1343
    Insert for 0,5 ml and 1,0 ml screw top tube caps
    Insert for 0,5 ml and 1,0 ml screw top tube caps
    White
    Log in for availability
    Log in for availability
    1 * 500 ST
    1 * 500 ST - 38,60 €
    38,60 €
    1 * 500 ST
       38,60 €
    Specifications:
      Low Manufacturing Impact
    More
    Specifications
    Cat. No.
    479-1343
    Environmentally Preferable
    Low Manufacturing Impact
    Environmental certification
    NAR Certificate of Retirement
      Low Manufacturing Impact