Order Entry
Belgium
ContactUsLinkComponent
Staining jar and basket
Staining jar and basket
Catalog # 631-9810
Supplier:  DELTALAB
Staining jar and basket
Catalog # 631-9810
Supplier:  DELTALAB

Specifications

  • Description:
    Staining jar with lid, adjustable
  • Seq:
    01
  • W×D×H:
    115×88×77 mm
  • Pk:
    1

Specifications

About this item

Made of stainless steel, with lid and adjustable handle.

  • With lid to avoid evaporation
  • Excellent chemical resistance
  • Excellent temperature resistance

The staining basket hold up to 30 slides (76×26 mm).