GL-Aeration for bottles, BOLA
Supplier: Bohlender
Certificates
About this item
PPS screw cap and PTFE body.
- Chemical resistant
- Temperature resistant from –40 to +70 °C
Filter with PTFE membrane (pore size 0,2 µm) and flexible tubing, screw cap with GL threads. Application: For sterile pressure compensation on multiple distributors for bottles. Overpressure or vacuum in the vessel are prevented. A contamination of the product with dust or any other particles is avoided.
Caution: It is recommended to exchange the filter every six months.
Frequently Bought Together