Order Entry
Belgium
ContactUsLinkComponent
Screw caps with aperture, BOLA
Screw caps with aperture, BOLA
Catalog # BOHLH989-40
Supplier:  Bohlender
undefined
Screw caps with aperture, BOLA
Catalog # BOHLH989-40
Supplier:  Bohlender

Specifications

  • Thread:
    S 40
  • Colour:
    Anthracite
  • Pk:
    1

Specifications

About this item

Screw cap, made of PPS, with handy knurl, with aperture.

  • Diameter of aperture: 28 mm
  • Universal chemical resistance
  • Temperature resistance: from –20 to +250 °C
  • Suitable gasket rings are available separately

As joining piece between tubes with flange and tubes with GL thread.