Orders LinkContactUsLinkComponent
Adapters
Adapters
Supplier:   VICI JOUR RESEARCH
Adapters
Supplier:  VICI JOUR RESEARCH
Certificates
10 Options Available Below
Product Details & Documents
These PEEK adapters connect standard LC fittings to standard LC ports, usable with most common organic and inorganic HPLC solvents without risking corrosion, metal contamination, or protein degradation.
  • Easy connection of solvent lines
  • Low dead volume
  • Completely inert and biocompatible
Pressure rating of M6, 1/4–28 is <70 bar (1000 psi), Luer is <35 bar (509 psi) and varies with tubing material and ID.
Product Family Options
PEEK adapters
Product Information
  • Hole Ø
    Availability
    Price
  • JOURJR-062
    Adapter, 1/4"-28 female to 10-32 female
    Adapter, 1/4"-28 female to 10-32 female
    0,40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 26,70 €
    26,70 €
    1 * 1 ST
       26,70 €
  • JOURJR-5505
    Adapter, M6 female to 10-32 male
    Adapter, M6 female to 10-32 male
    0,40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 30,20 €
    30,20 €
    1 * 1 ST
       30,20 €
  • 546-1383
    Adapter, 10-32 female to M6 female (Pharmacia)
    Adapter, 10-32 female to M6 female (Pharmacia)
    0,40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 27,60 €
    27,60 €
    1 * 1 ST
       27,60 €
PEEK port and Luer adapters
Product Information
  • Hole Ø
    Availability
    Price
  • 546-1251
    Adapter, Waters female to 10-32 male
    Adapter, Waters female to 10-32 male
    0,40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 36,40 €
    36,40 €
    1 * 1 ST
       36,40 €
  • 554-1003
    Adapter, 1/4"-28 female to 10-32 male
    Adapter, 1/4"-28 female to 10-32 male
    0,40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 92,80 €
    92,80 €
    1 * 1 ST
       92,80 €
  • 554-1008
    Adapter, 10-32 female to 1/4”-28 male
    Adapter, 10-32 female to 1/4”-28 male
    0,40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 92,80 €
    92,80 €
    1 * 1 ST
       92,80 €
  • 554-1013
    Adapter, 1/4"-28 female to M6 male
    Adapter, 1/4"-28 female to M6 male
    0,40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 92,80 €
    92,80 €
    1 * 1 ST
       92,80 €
  • 554-1014
    Adapter, 10-32 female to Luer female
    Adapter, 10-32 female to Luer female
    0,40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 73,60 €
    73,60 €
    1 * 1 ST
       73,60 €
  • 554-1016
    Adapter, 1/4"-28 female to Luer female
    Adapter, 1/4"-28 female to Luer female
    0,40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 53,00 €
    53,00 €
    1 * 1 ST
       53,00 €
  • 554-1017
    Adapter, 1/4" 28 female to Luer male
    Adapter, 1/4" 28 female to Luer male
    0,40 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 53,00 €
    53,00 €
    1 * 1 ST
       53,00 €