ContactUsLinkComponent
Flow through cooler CK 310
Flow through cooler CK 310
Catalog # SCOI285414320
Supplier:  SI Analytics
undefined
Flow through cooler CK 310
Catalog # SCOI285414320
Supplier:  SI Analytics
Specifications
  • Description:
    Flow-through cooler
  • Pk:
    1
  • Type:
    CK 310
Specifications
About this item
The flow-through cooler CK 310 is the perfect complement to bath thermostats of CT 72 series to extending their temperature range to below ambient temperature.
  • Small footprint
  • Low energy requirement: Economic solution preventing tap water usage for cooling
  • Internal cooling loop is made of stainless steel: Better chemical resistance than copper