Order Entry
Belgium
Orders LinkContactUsLinkComponent
Cell scrapers

Cell scrapers

Mini cell scraper, PE, disposable, fits 24, 48 and 96-well plates, blade width 5 mm, 60 mm
Catalog # 22003.
Supplier:   Biotium
Cell scrapers
Catalog # 22003.
Supplier:  Biotium
Cell scrapers
Cell scrapers

Mini cell scraper, PE, disposable, fits 24, 48 and 96-well plates, blade width 5 mm, 60 mm

Price
442,00 €
Log in for availability
1 * 200 ST
Product Details & Documents

PE, disposable, sterile. Mini cell scrapers for harvesting cells or cell lysates from microwell plates.

Packaging: Packaged in 20 sterile packs of 10 scrapers per pack.

Specifications
  • Description:
    Mini cell scraper, PE, disposable, fits 24, 48 and 96-well plates, blade width 5 mm
  • Total length:
    60 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.