Orders LinkContactUsLinkComponent
Flow cups with fixed nozzle
Flow cups with fixed nozzle
Flow cups with fixed nozzle
Flow cups with fixed nozzle
These flow cups are composed of an anodised aluminium cup and a stainless steel nozzle.
Designed to measure the viscosity of lacquers, paint and other liquids.
 : ISO 2431
 : Stand must be ordered separately.
Product Family Options
  • Description
    Range
    Cup orifice
  • Viscosity flow cup (max.W×H = 92×84 mm)
    7 - 42 cSt
    3 mm
    1 * 1 ST
    1 * 1 ST - 201,00 €
    201,00 €
    1 * 1 ST
       201,00 €
  • Viscosity flow cup (max.W×H = 92×84 mm)
    34 - 135 cSt
    4 mm
    1 * 1 ST
    1 * 1 ST - 201,00 €
    201,00 €
    1 * 1 ST
       201,00 €
  • Viscosity flow cup (max.W×H = 92×84 mm)
    91 - 326 cSt
    5 mm
    1 * 1 ST
    1 * 1 ST - 222,00 €
    222,00 €
    1 * 1 ST
       222,00 €
  • Viscosity flow cup (max.W×H = 92×84 mm)
    188 - 684 cSt
    6 mm
    1 * 1 ST
    1 * 1 ST - 201,00 €
    201,00 €
    1 * 1 ST
       201,00 €
  • Viscosity flow cup (max.W×H = 92×84 mm)
    600 - 2000 cSt
    8 mm
    1 * 1 ST
    1 * 1 ST - 222,00 €
    222,00 €
    1 * 1 ST
       222,00 €
Accessories
  •  
    Description
  • SIMEDI0076
    Digital stopwatch, 0,01 s
    1 * 1 ST - 29,30 €
  •  
    Description
  • SIMEVF2062
    Ring stand fro flow cups
    1 * 1 ST - 83,00 €
  •  
    Description
  • SIMEVF2068
    Temperature control jacket with tripod for ISO and AFNOR cups
    1 * 1 ST - 621,00 €
  •  
    Description
  • 769-1151
    Viscosity convertor of flow time to viscosity
    1 * 1 ST - 86,60 €