Orders LinkContactUsLinkComponent
Staining plates
Staining plates
Staining plates
Certificates
Staining plates
2 Options Available Below
Product Details & Documents
Glass spot plates with 10 or 12 numbered cavities. For flocculation tests.
Product Family Options
Product Information
  • No. of wells
    Well size
    W×D×H
    Availability
    Price
  • 10
    Ø×D: 14×2 mm
    105×55×5 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 37,40 €
    37,40 €
    1 * 1 ST
       37,40 €
  • 12
    Ø×D: 15×0,8 mm
    89×57×4 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 40,60 €
    40,60 €
    1 * 1 ST
       40,60 €