BioClean-C™ Chemo Protective Aprons with Sleeves
: Nitritex
These disposable aprons are made of CleanTough™ fabric, a spunbonded non woven PP fabric laminated with a PE film. The elasticated cuffs are made of 100% polyester.
- Processed to ensure ISO Class 4 compatibility
- Lightweight, low linting and hard-wearing CleanTough™ fabric
- Ultrasonically bonded seams with protective tape
- Adjustable neck fastening
- Tie tapes at the waist
Provide protection against fine sprays and particles as well as against a range of chemotherapy drugs.
Colour: Blue.
: EN ISO 13034:2005 + A1:2010 (Type PB [6]), EN 14325, EN 530, EN 6530, EN 13934-1, EN 9073-4, EN 863, EN 13935-2; Cat. III
Manufactured in a facility holding ISO 9001:2015 and (EU) 2016/425 Module D certifications.
Manufactured in a facility holding ISO 9001:2015 and (EU) 2016/425 Module D certifications.
: One sealed piece per sealed inner bag; one sealed inner bag per sealed outer bag; 40 outer bags per carton liner; one carton liner per carton (40 pieces).
- Catalog No:
- NITRBCAS-XL
- 113-8146
- 113-8163
- 113-8164
- 113-8147
- 113-8162
- 113-8148
- Fabric:
- CleanTough™ (PP/PE)
- CleanTough™ (PP/PE)
- CleanTough™ (PP/PE)
- CleanTough™ (PP/PE)
- CleanTough™ (PP/PE)
- CleanTough™ (PP/PE)
- CleanTough™ (PP/PE)
- Cleanroom class:
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- 10/ISO 4
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Garment application:
- Cleanroom, Medical, Pharmaceutical, Clinical
- Cleanroom, Medical, Pharmaceutical, Clinical
- Cleanroom, Medical, Pharmaceutical, Clinical
- Cleanroom, Medical, Pharmaceutical, Clinical
- Cleanroom, Medical, Pharmaceutical, Clinical
- Cleanroom, Medical, Pharmaceutical, Clinical
- Cleanroom, Medical, Pharmaceutical, Clinical
Frequently Bought Together