ContactUsLinkComponent
Sampling spoons, wide handle, eco-friendly, SP Bel-Art
Sampling spoons, wide handle, eco-friendly, SP Bel-Art
Catalog # BELAF367401025
   Waste Reducing
undefined
Sampling spoons, wide handle, eco-friendly, SP Bel-Art
Catalog # BELAF367401025
   Waste Reducing
Specifications
  • Capacity:
    20 ml
  • Seq:
    05
  • Environmentally Preferable:
    Waste Reducing
  • Pk:
    10
Specifications
About this item
PLA (polylactic acid) resin, eco-friendly, transparent.
  • Green alternative to petroleum-based sampling tools; helps meet sustainability goals
  • FDA grade spoons are safe for food, drug and cosmetic contact
  • Comparable in strength and durability to PS, PP and other plastic resin sampling tools
  • Perfect for retrieving small, specific volume samples of powders or liquids
  • Wide handles for firm grip, provide just enough stiffness to easily sample packed powders and other firm materials
  • Unique flat bottom shape allows convenient duo use as weighing boats to save time and assure accuracy
The production of 100% plant-based PLA contributes less greenhouse gasses and uses less non renewable energy from source to resin than its petroleum-based counterparts. Disposal options include industrial composting for biodegradability, incineration, recycling and landfill. Use a separate spoon for each material sampled to avoid cross contamination.