Orders LinkContactUsLinkComponent
Squared bossheads
Squared bossheads
Supplier:   BOCHEM
Squared bossheads
Supplier:  BOCHEM
Certificates
Squared bossheads
Squared bossheads
2 Options Available Below
Product Details & Documents
Squared bossheads with hexagon socket screws.
  • Made from 18/10 stainless steel or aluminium (laboral)
  • Angle: 90°
Product Family Options
Product Information
  • Material
    For rod Ø
    Availability
    Price
  • Stainless steel 18/10
    12 - 13 × 12 - 13 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 50,30 €
    50,30 €
    1 * 1 ST
       50,30 €
  • Aluminium alloy
    12 - 13 × 12 - 13 mm
    Log in for availability
    Log in for availability
    1 * 1 ST
    1 * 1 ST - 22,90 €
    22,90 €
    1 * 1 ST
       22,90 €