Order Entry
Belgium
ContactUsLinkComponent
Tetrabutylammonium hydroxide 0.1 mol/l (0.1 N) in 2-propanol 66.6% (v/v)/methanol 33.3% (v/v)
Flammable
Corrosive
Health
Irritant
Tetrabutylammonium hydroxide 0.1 mol/l (0.1 N) in 2-propanol 66.6% (v/v)/methanol 33.3% (v/v)
Catalog # CMPLFC305940
CAS Number:  
Flammable
Corrosive
Health
Irritant
Tetrabutylammonium hydroxide 0.1 mol/l (0.1 N) in 2-propanol 66.6% (v/v)/methanol 33.3% (v/v)
Catalog # CMPLFC305940
CAS Number:  

Specifications

  • Pack type:
    Plastic bottle
  • Pk:
    1 L
  • Seq:
    5037

Specifications

About this item

 :
 :259.48 g/mol
Storage Temperature:Refrigerator
EINECS:218-147-6
UN:1992
ADR:3,II
 :GHS02
 :GHS05
 :GHS08
 :GHS07
CAS Number:2052-49-5
Formula:C₁₆H₃₇NO
MDL Number:MFCD00009425