Order Entry
Belgium
ContactUsLinkComponent
1-Ethyl-3-methylimidazoliumbis(trifluoromethylsulphonyl)imide ≥98.0% (by HPLC, titration analysis)
Irritant
1-Ethyl-3-methylimidazoliumbis(trifluoromethylsulphonyl)imide ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI
Certificates
undefined
Irritant
1-Ethyl-3-methylimidazoliumbis(trifluoromethylsulphonyl)imide ≥98.0% (by HPLC, titration analysis)
Supplier:  TCI

Specification Test Results

  • Appearance:
    Colorless to Light orange to Yellow clear liquid
  • Purity (HPLC):
    min. 98.0 area%
  • Purity (Ion exchange titration):
    min. 98.0 %

Specification Test Results

Related Information

Frequently Bought Together