
Fast green FCF
Supplier: Cayman Chemical
Certificates
About this item
CAS Number:2353-45-9
:
Formula:C₃₇H₃₄N₂Na₂O₁₀S₃
MDL Number:MFCD00013053
:808.86 g/mol
Merck Index:13,03970
Melting Pt:290 °C
EINECS:219-091-5
:GHS08
Storage Temperature:Ambient
Frequently Bought Together