Kjeldahl catalyst acc. to Wieninger
Catalog # 1.08030.1000
Supplier: Merck
CAS Number:
Specifications
- Pk:1 kg
- Description:Kjeldahl catalyst acc. to Wieninger
- Environmentally Preferable:
- Contents:>94% Na₂SO₄; >1,45% CuSO₄; >1,5% Se
Specifications
About this item
Packaging: Plastic bottle for solids.