Guaifenesin ≥98.0% (by HPLC and UV method)
Supplier: TCI
Certificates
About this item
Synonyms: Guaiacol glyceryl ether, 3-(2-Methoxyphenoxy)-1,2-propanediol, 3-(2-Methoxyphenoxy)propane-1,2-diol
CAS Number:93-14-1
:
Formula:C₁₀H₁₄O₄
MDL Number:MFCD00016873
:198.22 g/mol
Merck Index:14,04555
EINECS:202-222-5
:GHS07
Specifications
- Environmentally Preferable:
Specifications
Specification Test Results
- Appearance:White powder to crystal
- Purity (HPLC):min. 98.0 area%
- Purity (UV-vis method):98.0 to 102.0 %
- Melting point:80.0 to 84.0 deg-C
- Solubility in hot Water:almost transparency
Specification Test Results
Frequently Bought Together